Learn More
DuPont™ Tychem™ 2000 SFR Hooded Coverall
Catalog No. 18888722 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-722 3X-Large
18-888-721 2X-Large
18-888-723 4X-Large
18-888-724 5X-Large
18-888-725 Large
18-888-726 Medium
18-888-727 Small
18-888-728 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-722 Supplier DuPont™ Supplier No. QS128TGR3X000400
Add to Cart
Add to Cart
This item is not returnable. View return policy
TYCHEM™ 2000 SFR Coverall features chemical and secondary flame resistance
  • Respirator-fit hood lined with ProShield™ 6 SFR fabric.
  • Manufactured under specifications that do not contain natural rubber latex
  • Covered, braided elastic at hood.
  • Nylon zipper with large metal pull
  • Chin flap with double-sided adhesive tape
  • Storm flap covers zipper which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper.
  • Covered, braided elastic openings at wrist for tighter fit.
  • Attached socks composed of garment material.
  • Outer boot flaps with elastic are designed to cover boot tops to help reduce potential for liquid intrusion.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Gender Unisex
Material Tychem
Color Olive Green
Disposable Single-use
Ankle Style Attached Socks, Boot Overflaps, Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Flame, Acid
Latex Free Yes
No. of Pockets None
Quantity 4/Pk.