Learn More
DuPont™ Tychem™ 4000 Coverall (all)
Catalog No. 17500133 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
6X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-133 3X-Large
17-500-134 4X-Large
17-500-135 5X-Large
17-500-136 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-500-133 Supplier DuPont™ Supplier No. SL120BWH3X001200
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ TYCHEM™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals, Bloodborne Pathogens
No. of Pockets None