Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 189992217 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-2217 3X-Large
01-361-51B Medium
01-361-51C Large
01-361-51D X-Large
18-999-2216 2X-Large
19-166-136 4X-Large
19-088-1701 5X-Large
17-500-147 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-999-2217 Supplier DuPont™ Supplier No. SL122BWH3X001200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls with Socks
Size 3X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Bound
Sterility Non-sterile
Description Attached hood, elastic wrist, attached socks
Size (English) Chest 47.25 to 50.75 in.
Size (English) Inseam 31 in.
Size (Metric) Inseam 78.74 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Width (English) Chest 30.25 in.
Width (Metric) Chest 76.83 cm
Height (English) 6.16 to 6.33 ft.
Height (Metric) 1.82 to 1.92 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.