Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Catalog No. 18999593 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-593 5X-Large
19-120-694B Medium
18-643 Large
18-642 X-Large
18-641 2X-Large
19-120-694C 3X-Large
18-999-592 4X-Large
17-500-151 7X-Large
17-500-150 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 18-999-593 Supplier DuPont™ Supplier No. SL127BWH5X001200
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Size 5X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals, Bloodborne Pathogens
No. of Pockets None