Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 19715334 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Seam Type:
Bound
Taped
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19-715-334 Medium Taped
19-162-778 5X-Large Taped
19-715-335 Large Taped
18-999-2583 4X-Large Bound
18-760D X-Large Bound
18-760E 3X-Large Bound
19-162-777 4X-Large Taped
19-715-336 2X-Large Taped
18-760C Large Bound
19-087309 2X-Large Bound
18-760B Medium Bound
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-715-334 Supplier DuPont™ Supplier No. SL125TWHMD000600
Add to Cart
Add to Cart
This item is not returnable. View return policy

Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Gender Unisex
Material Tyvek
Color White
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Seam Type Taped
Coating Saranex 23-P film
Size (English) Chest 35.25 to 38.75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Chest 89.53 to 98.42 cm
Size (Metric) Inseam 69.85 cm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.