Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
Catalog No. 17500042 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
3X-Large
4X-Large
5X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-042 3X-Large
17-500-044 4X-Large
17-500-046 5X-Large
17-500-049 Medium
17-500-051 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-500-042 Supplier DuPont™ Supplier No. C3128TTN3X000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 5000 Coverall with Respirator Fit Hood, Attached Socks provides protection against a broad range of chemicals and situations involving liquid splash.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Gender Unisex
Material Tychem
Color Tan
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Closure Type Zipper
Seam Type Taped
Style Respirator Fit Hood