Description
- Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Strong zipper for tough, flexible closure
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
| Size | 4X-Large |
| Gender | Unisex |
| Material | Tychem |
| Color | Tan |
| Ankle Style | Attached Socks, Boot Overflaps |
| Cuff Style | Elastic |
| Closure Type | Zipper |
| Seam Type | Taped |
| Style | Respirator Fit Hood |