Learn More
DuPont™ Tychem™ 5000 Coverall with Respirator Hood, Berry Compliant
Shop All DuPont Personal Protection ProductsDescription
DuPont™ Tychem™ 5000 helps offer protection for those in hazardous environments.
- Provides at least 30 minutes of protection against over 150 chemical challenges
- NFPA 1992 certified
Compliance
BN=BERRY COMPLIANT
Specifications
Specifications
| Size | 5X-Large |
| Color | Tan |
| Ankle Style | Attached Socks, Boot Overflaps |
| Cuff Style | Elastic |
| Closure Type | Storm Flap with Adhesive Closure |
| Seam Type | Taped |
| Style | Respirator Fit Hood |