Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 5000 Coveralls with Respirator Fit Hoods, Berry Amendment Compliant

Catalog No. 19715275 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Certifications/Compliance:
ASTM D1777
Closure Type:
Zipper with Storm Flap
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Closure Type Size Height (English)
19-715-275 ASTM D1777 Zipper with Storm Flap 4X-Large 6.4 to 6.7 ft.
19-715-270 ASTM D1777 Zipper with Storm Flap Medium 5.3 to 5.7 ft.
19-715-271 ASTM D1777 Zipper with Storm Flap Large 5.5 to 5.9 ft.
19-715-274 ASTM D1777 Zipper with Storm Flap 3X-Large 6.2 to 6.4 ft.
19-715-272 ASTM D1777 Zipper with Storm Flap X-Large 5.8 to 6.2 ft.
19-715-273 ASTM D1777 Zipper with Storm Flap 2X-Large 6 to 6.4 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-715-275 Supplier DuPont™ Supplier No. C3128TTN4X0006BN
This item is not returnable. View return policy

Designed for use during rigorous activities

Coveralls protect against chemical splash as well as potential exposure to chemical warfare agents. Tan-colored polypropylene substrate offers greater physical strength and chemical holdout.

  • Multilayer barrier film laminated to a lasting 2 oz. per sq. yd.
  • Respirator-fit attached hood,
  • Front zipper closure with double storm flaps
  • Elastic wrists
  • Outer boot flaps

Compliance

Meets ASTM™ D1777 standards.

†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

TRUSTED_SUSTAINABILITY

Specifications

Size 4X-Large
Material Tychem
Color Tan
Disposable Single-use
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Zipper with Storm Flap
Seam Type Taped
Certifications/Compliance ASTM D1777
Description Tychem™ CPF 3 a multilayer composite barrier film laminated to a durable 2 oz. Polypropylene substrate
Size (English) Chest 55.25 to 58.75 in.
Size (English) Inseam 31.75 in.
Size (Metric) Chest 140.33 to 149.22 cm
Size (Metric) Inseam 80.64 cm
Size (Metric) Waist 158.75 cm
Thickness 0.45 mm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 40.25 in.
Length (Metric) Sleeve 102.2 cm
Width (English) Chest 34.25 in.
Height (English) 6.4 to 6.7 ft.
Height (Metric) 1.95 to 2.04 m
Weight (English) 0.27 lb.
Weight (Metric) 0.12 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.