Description
- Ideal for applications that do not require the full body coverage of a coverall.
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Mandarin collar designed to fit tightly around hooded PAPR
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Comes with a jam fit cuff and glove insert for user supplied gloves
- Typically worn with C3360T TN pants or sold as jacket/pant ensemble (C3750T TN)
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
| Size | Large |
| Gender | Unisex |
| Material | Tychem |
| Color | Tan |
| Closure Type | Zipper |
| Collar Type | Mandarin Collar |
| Cuff Style | Jam Fit |
| Seam Type | Taped |
| Style | Extends to Hip |