Learn More
DuPont™ Tychem™ 6000 Coverall with Attached Hood, Socks, Butyl Gloves
Catalog No. 18888791 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-791 Medium
18-888-788 4X-Large
18-888-789 5X-Large
18-888-792 Small
18-888-786 2X-Large
18-888-787 3X-Large
18-888-790 Large
18-888-793 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-791 Supplier DuPont™ Supplier No. TF186TGYMD0006PI
Add to Cart
Add to Cart
This item is not returnable. View return policy
Coveralls provide protection from chemical warfare agents and toxic industrial chemicals.
Tychem™ 6000 coveralls are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Material Tychem
Size Medium
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Description Packaged Individually
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Quantity 6 Pk.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile