Learn More
DuPont™ Tychem™ 6000 FR Hooded Coverall
Catalog No. 18888812 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Gray
Orange
Size:
Small
5X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-888-812 Orange Small
18-888-810 Gray 5X-Large
18-888-811 Gray Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-812 Supplier DuPont™ Supplier No. TP198TORSM000200
This item is not returnable. View return policy
Tychem™ 6000 FR offers triple-hazard protection from liquid-chemical splash, flash fire, and electric arc.
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached respirator-fit hood with drawstring to pull tightly around respirator; longer zipper extends to chin for complete coverage of neck area.
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Open ankles, hemmed
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Certifications
NFPA 1992 and 2112
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Gender Unisex
Material Tychem
Color Orange
Disposable Single-use
Ankle Style Open
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Liquid
Arc Flash Rating 15 cal/cm2
Certifications/Compliance NFPA 1992 and 2112
Latex Free Yes
No. of Pockets None
Quantity 2/Pk.