Learn More
DuPont™ Tychem™ 9000 Coverall with Respirator Fit Hood, Berry Compliant
Catalog No. 17500005 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-005 2X-Large
17-500-006 3X-Large
17-500-007 4X-Large
17-500-008 5X-Large
17-500-009 Large
17-500-010 Medium
17-500-012 Small
17-500-013 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-500-005 Supplier DuPont™ missing translation for 'supplierNo' BR127TYL2X0002BN
This item is not returnable. View return policy
DuPont™ Tychem™ 9000 Coverall with Respirator Fit Hood, Berry Compliant, offers protection against a wide range of chemical challenges, resistance to tears, punctures, and abrasion.
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
  • NFPA 1994 Class 2 and NFPA 1992 certified
  • Provides at least 30 minutes of protection against over 280 chemical challenges
  • Used by many in the chemical handling and petrochemical manufacturing
Compliance
BN=BERRY COMPLIANT
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Color Yellow
Ankle Style Elastic
Cuff Style Elastic
Closure Type Storm Flap with Adhesive Closure
Seam Type Taped
Style Respirator Fit Hood