Learn More
DuPont™ Tychem™ 9000 Coverall with Respirator Fit Hood, Berry Compliant
Shop All DuPont Personal Protection ProductsDescription
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
- NFPA 1994 Class 2 and NFPA 1992 certified
- Provides at least 30 minutes of protection against over 280 chemical challenges
- Used by many in the chemical handling and petrochemical manufacturing
Compliance
BN=BERRY COMPLIANT
Specifications
Specifications
| Size | Small |
| Color | Yellow |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Closure Type | Storm Flap with Adhesive Closure |
| Seam Type | Taped |
| Style | Respirator Fit Hood |