Description
- Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
- Also provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
- Standard 20 mil PVC faceshield
- Rear entry design keeps closure away from direct contact with frontal chemical exposure
- Flat back design for use with airline
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Specifications
Specifications
| Color | Tan |
| Material | Tychem |
| Size | 2X-Large |
| Ankle Style | Attached Socks And Boot Flaps |
| Closure Type | Zipper |
| Seam Type | Taped |