Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ NP560 Chemical Resistant Gloves, Seamless Knit Liner

Catalog No. 19715415 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
9
10
11
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-715-415 11
19-715-413 9
19-715-414 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-715-415 Supplier DuPont™ Supplier No. NP5600BK110144KL
This item is not returnable. View return policy

Flexible neoprene gloves are resistant to a wide range of chemicals, including acids, caustics, solvents, greases and oils

  • Flexible, consistent elastomeric coating
  • Tactile non-slip grip
  • 15-gauge seamless knit liner for maximum comfort
  • Recommended for mechanical, petrochemical and automotive markets
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) Cat III CE 2777
Color Black
Hand Specific
Latex Free Yes
Material Neoprene
Size 11
Thickness (English) 55 mil
Thickness (Metric) 1.39 mm
EN388 Standard EN 388:3031X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Rough
Liner Material Seamless Knit Polyester
Lining Lined
Model NP5600BK110144KL
Series Tychem™ NP560
Length (English) 14 in
Length (Metric) 35.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.