Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ NT450 14 in. Lined Gloves

Catalog No. 19715391 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
8
9
10
11
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-715-391 8
19-715-392 9
19-715-393 10
19-715-394 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-715-391 Supplier DuPont™ Supplier No. NT4500BU080144KL
This item is not returnable. View return policy

Flexible impermeable cotton-polyester lined dual nitrile glove. Ideal for heavy duty work or longer exposure to hazardous chemicals and solvents.

  • Protects against an array of solvents, oils and acids
  • Cotton liner absorbs perspiration for comfortable extended wear, with good mechanical resistance
  • Protects the hand and forearm from oils, hydrocarbons, grease and abrasion
  • Double nitrile coating
  • Impermeable for working in damp or greasy environments.
  • Rough textured finish over surface of hand
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) Cat III CE 2777
Chlorinated No
Color Blue
Disposable Single-use
Exterior Finish Rough
Hand Specific
Latex Free Yes
Material Nitrile
Size 8
Thickness (English) 56 mil
Thickness (Metric) 1.42 mm
EN388 Standard EN 388:4112X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Scalloped
Grip Design Rough
Liner Material Cotton/Polyester
Lining Lined
Model NT4500BU080144KL
Series Tychem™ NT450
Length (English) 14 in.
Length (Metric) 35.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.