Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with Hansen Fitting
Shop All DuPont Personal Protection ProductsDescription
- CSM Encapsulated Level A Suit with Hansen Fitting design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
- Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing
- Standard-width, anti-fog, three-layer (PVC 40 mil / Teflon™ 5 mil / PVC 20 mil) faceshield
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Specifications
| Color | Tan |
| Material | Tychem |
| Size | 3X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Seam Type | Taped |