Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with Interspiro
Catalog No. 18888735 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-735 3X-Large
18-888-732 2X-Large
18-888-738 4X-Large
18-888-743 Large
18-888-746 Medium
18-888-749 Small
18-888-752 X-Large
18-888-754 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-735 Supplier DuPont™ Supplier No. RC550TTN3X00017W
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
CSM Encapsulated Level A Suit is a front entry, low visibility suit with attached socks and gloves
  • CSM Encapsulated Level A Suit design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing
  • Standard-width, anti-fog, three-layer (PVC 40 mil / Teflon™ 5 mil / PVC 20 mil) faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
TRUSTED_SUSTAINABILITY
Specifications
Color Tan
Material Tychem
Size 3X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped