Learn More
Saint Gobain Performance Plastics Tube Tygon .010 ID 500FT-150M/PK
Supplier:  Saint Gobain Performance Plastics AAD04091
Encompass_Preferred
Saint-Gobain Tygon ND 00-80 Tubing I.D. 0.01 in. (0.25mm) O.D 0.03 in. (0.76mm) 0.01 in. (0.25mm) thick Needle gauge 30 Diameter Inner 0.01 in. Diameter Outer 0.03 in. Thickness 0.01 in. Clear For Use With Medical grade
Catalog No. 50-206-8920
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.