Learn More
ATCC TYGM-9 Medium, 8ml
Supplier:  ATCC PRA1171
Encompass
This medium is the recommended formulation for the xenic (ie, bacterized) cultivation of Entamoeba, Dientamoeba, and Endolimax. It is provided in liquid form (8 mL), complete with 3% bovine serum; ready to use.
Catalog No. 50-238-3826
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.