Learn More
Saint Gobain Performance Plastics TYGN SILVER ANTMCROBIAL 50/PK
Supplier:  Saint Gobain Performance Plastics AS600012
Encompass_Preferred
Saint-Gobain Tygon Silver Antimicrobial Tubing Diameter Inner 4.8 mm 3/16 in. Diameter Outer 7.9 mm 5/16 in. Plastic Overall Length 50 ft. 15 m Thickness 1/16 in. 1.6 mm
Catalog No. 50-206-9032
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.