Learn More
Saint Gobain Performance Plastics TYGOPRENE x L-60 PUMP TUBING
Supplier:  Saint Gobain Performance Plastics AN800007
Encompass_Preferred
Saint-Gobain Tygon Peristaltic Pump Tubing XL-60 Diameter Inner 0.13 in. Diameter Outer 1/4 in. Tygoprene Thickness 0.0625 in. Maximum Pressure 20 psi Must order minimum quantity of 2.
Catalog No. 50-206-8902
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.