Learn More
Cayman Chemical BMY 14802hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 4226410 MG
Encompass_Preferred
A Cdk1 and Cdk2 inhibitor (IC50 0.002 UM for both) inhibits LoVo cell proliferation (IC50 0.039 UM) reduces tumor volume in an SW620 colon adenocarcinoma mouse xenograft model at 15 mg/kg
Catalog No. 50-294-3284
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.