Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Tylenol™ Extra Strength Tablets

Catalog No. 19090829 Shop All Medique Products
Change view
Click to view available options
Quantity:
50 x 2/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-829 50 x 2/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-829 Supplier Medique Supplier No. 49733
Only null left
Add to Cart
Edge
Add to Cart

Pain reliever and fever reducer tablets.

  • Each tablet contains: Acetaminophen 500mg

Specifications

Active Ingredients Acetaminophen 500 mg
Product Type Tylenol™ Tablets
For Use With (Application) Pain reliever and fever reducer
Format Extra Strength
Quantity 50 x 2/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.