Learn More
Abcam Tylo x apol, 5G
Supplier:  Abcam AB2854295G
Encompass_Preferred
MW 280.4 Da. A 10% solution of the nonionic liquid surfactant Tyloxapol in water. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.