Learn More
International Crystal UV-VIS Cells/Type 1 Standard Cell with 10mm lightpath, 'Q'
Supplier:  International Crystal 0023Q236
Encompass_Preferred
ICL cells are completely fused without the use of low melting intermediate glasses or cements. In other words, quartz is fused to quartz and glass is fused to glass. This eliminates strains due to differences in coefficients of expansion found in cells fused with intermediate low melting glasses. Because of the fused construction, the window joints are unaffected by all types of cleaning solutions.
Catalog No. NC1408383
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.