Learn More
TECHREF™ Type AT-700 Assembly Tool
SDP
Catalog No. 11022105 Shop All TECHREF Inc. Products
Change view
Click to view available options
Type:
Assembly Tool
Catalog No. Type
11-022-105 Assembly Tool
1 options
Catalog No. 11-022-105 Supplier TECHREF™ Supplier No. AT700
Add to Cart
Add to Cart
Designed and built to ease the assebmley of the TECHREF Type C-700 sample cells.
Use the AT-700 tool to seat the cell ring down firmly over the cup to create a flat, tight and wrinkle-free film surface.
  • Made of aluminum and anodized with a fluoropolymer coating
Specifications
Type Assembly Tool
Material Aluminum and anodized with a fluoropolymer
For Use With (Equipment) TECHREF Type C-700 Sample Cells
WARNING: Cancer - www.P65Warnings.ca.gov