Learn More
Leica Microsystems Type F Immersion Liquid, ISO 8036
Catalog No. 16800491 Shop All Leica Microsystems Products
Change view
Click to view available options
Type:
Immersion Oil
Catalog No. Type
16-800-491 Immersion Oil
1 options
Catalog No. 16-800-491 Supplier Leica Microsystems Supplier No. 11513859
Add to Cart
Add to Cart
Type F Immersion liquid for oil immersion microscope objective lenses.
  • Type F Immersion liquid, ISO 8036
  • 10 ml
  • Very low autofluorescence
  • Highly recommended for fluorescence applications and all APO objectives
TRUSTED_SUSTAINABILITY
Specifications
Type Immersion Oil
Viscosity Low