Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Werner Type IA Aluminum Step Stool

Step stool that is ideal for plant, office or home applications The molded top provides a large standing platform.

Supplier:  Werner 150B

Catalog No. 17-010-755


Only null left
Add to Cart

Description

Description

Molded top provides large standing platform.

  • External Pinch-Proof™ spreaders
  • Slip-resistant Traction-Tred™ steps
  • Molded vinyl foot pads riveted to rails
  • Bottom step has durable aluminum braces
  • Ideal for construction, plant, office and home applications
  • Duty rating 300 lb.
  • Approx. shipping weight is 6 lb.
Specifications

Specifications

ANSI-A14-2-2007, OSHA
24 in.
6 ft.
17 in.
136.1 kg
Step Stool
24 x 17 x 6 in.
1 Ea.
150B Series
300 lb.
Aluminum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.