Learn More
Typenex™ Flexi Band Forms™
SDP
Catalog No. 22355216
Change view
Click to view available options
Product Type:
Condensed Patient ID Form
General Patient ID Form
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-355-216 Condensed Patient ID Form
22-355-215 General Patient ID Form
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-355-216 Supplier Typenex™ Supplier No. FLX002
Provide perforated inserts for Flexi bands
Choose from the general or condensed form, based on the number of stickers required
  • With barcoded blood bank ID numbers (BBIDs) for easy scanning into Laboratory Information System (LIS)
  • Inserts can be used for handwriting or with patient labels
  • Forms have 7-digit BBID in GS1-128 barcode format
  • General form provides two patient inserts and 18 barcoded specimen tube stickers
  • Condensed form is used for facilities that generate specimen tube stickers as needed; five stickers provided
Specifications
Product Type Condensed Patient ID Form
Includes 5 Stickers, 7-digit BBID in GS1-128 barcode format
For Use With (Equipment) Flexi bands
Barcode Yes