Learn More
Typenex™ SmearSafe™ Blood Dispenser
SDP
Catalog No. 22250937
Change view
Click to view available options
Blood dispenser
Catalog No.
22-250-937 Blood dispenser
1 options
Catalog No. 22-250-937 Supplier Typenex™ Supplier No. C1000
Add to Cart
Add to Cart
For easy dispensing of a drop of blood from a tube without removing the stopper
  • Fewer steps, less time when compared to use of applicator sticks or capillary tubes
  • Dispenses consistent drop size for uniform smears
  • Comfortable on fingers
  • Single-use device
  • Compatible with Vacutainer™ and Vacuette™ tubes
Specifications
Quantity 1000 Cs.
Type Blood Dispenser
For Use With (Application) For easy dispensing of a drop of blood from a tube without removing the stopper