Learn More
Cayman Chemical TyphnosIde 10mg
SDP
Supplier:  Cayman Chemical 3897010 MG
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; decreases colony size and induces ferroptosis and cell cycle arrest at the G2/M phase in Kasumi-1 and NB4 cells at 40 µM; reduces LPS-induced decreases in cell viability, increases in MDA and NO, and decreases in SOD activity in HUVECs at 70 µM; limits 4-aminopyridine-induced increases in glutamate secretion and intracellular calcium levels in isolated rat cerebral synaptosomes at 20 µM; typhaneoside, in complex with a tFNA, decreases the area and severity of necrotic kidney tissue, apoptotic kidney cells, blood urea nitrogen and serum creatinine levels, and increases mitochondrial membrane integrity in isolated kidney cells in a mouse model of renal ischemia-reperfusion injury induced by bilateral renal pedicle clamping
Catalog No. 50-291-7307
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.