Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Typical Flower Model
SCI_ED

Catalog No. S80810 Shop All Eisco Products
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S80810 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80810 Supplier Eisco™ Supplier No. BM0004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Depicts internal structures, including reproductive parts of a typical flower

  • Typical flower model for biological study
  • Three dimensional details depict internal structures
  • Numbered with key card for identification of features
  • Mounted on a base for display
  • Hand painted
  • Perfect for any classroom

Specifications

Color Hand Painted
Dimensions (L x W x H) 10.5 x 8.25 x 1.5 in. (Excluding Base)
Dimensions (L x W) 13 x 9 in. (Base)
For Use With (Application) Teaching The Reproductive Parts of a Typical Flower
Grade 9 to 12
Height (English) 2.5 in.
Height (Metric) 63.5 mm
Includes Numbered key card for structure identification
Material Plastic
Mounting Base
Quantity 1
Gender Male, Female
Product Type Botany Model
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.