Learn More
CPC Scientific H-Tyr-Ile-Tyr-Gly-Ser-Phe-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP051A
Encompass
Peptide selected from a random combinatorial peptide library that is also a much more specific and efficient substrate for the Src-family protein tyrosine kinase than cdc2 (6-20). Its Km value for p60 c-src (55 M) is about 6.4-fold lower than that of the cdc2 (6-20) substrate.SEQUENCE: H-Tyr-Ile-Tyr-Gly-Ser-Phe-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: YIYGSFKMOLECULAR FORMULA: C44H60N8O11MOLECULAR WEIGHT: 877.01STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [165190-42-1]SYNONYMS: RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): PKCP-051A
Catalog No. 50-211-0207