Learn More
CPC Scientific H-Tyr-Lys-Val-Gln-Asp-Asp-Thr-Lys-Thr-Leu-Ile-Lys-Thr-Ile-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific LEPT002A
Encompass
ONE-LETTER SEQUENCE: YKVQDDTKTLIKTIVMOLECULAR FORMULA: C80H137N19O25MOLECULAR WEIGHT:1765.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: ObesityREFERENCES: Y. Zhang et al., Nature, 372, 425 (1994); M.A. Pelleymounter et al., Science 269, 541 (1995)
Catalog No. 50-210-9633
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.