Learn More
Bachem (Tyr27)-a-CGRP (27-37) (canine, mouse, rat)
SDP
Supplier:  Bachem 4037183.0001
Encompass_Preferred
(Tyr27)-a-CGRP (27-37) (canine, mouse, rat), H-Tyr-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH2, 124501-79-7, C54H79N13O17, Mr 1182.30, 1mg. [Tyr0] Calcitonin Gene Related Peptide (28-37), rat.
Catalog No. 50-259-941