Learn More
CPC Scientific H-Ala-Phe-Leu-Pro-Trp-His-Arg-Leu-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP053B
Encompass
Tyrosinase epitope that is recognized by HLA-A24 restricted, tumor-infiltrating lyphocytes. It may be used as a reagent to generate melanoma-specific T cells for adoptive immunotherapy in addition to peptide vaccines for HLA-A24+ melanoma patients. ONE-LETTER SEQUENCE: AFLPWHRLFMOLECULAR FORMULA: C61H83N15O10MOLECULAR WEIGHT:1186.43STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [166188-11-0], SYNONYMS: Tyrosinase (206-214) (human)RESEARCH AREA: Cancer
Catalog No. 50-211-0211
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.