Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Tyrosine Decarboxylase (apoenzyme) (S. faecalis)

Catalog No. 89966299 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-966-299 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-966-299 Supplier Abnova™ Supplier No. P5293
Add to Cart
Add to Cart

Used for Enzyme

Tyrosine decarboxylase catalyzes the removal of the carboxyl group from tyrosine to produce tyramine and carbon dioxide.

Specifications

For Use With (Application) Enzyme Activity
Name Tyrosine Decarboxylase (apoenzyme) (S. faecalis)
Preparation Method Prepared by grown the cells in vitamin B6 deficient media and dried
Quantity 250 mg
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name TYROSINE
Biological Activity Activates to at least 0.2 units per mg. One Unit yields 1μmole of CO2 per minute from L-tyrosine at 37°C, pH 5.5. The apoenzyme activity is measured in the presence of excess pyridoxal phosphate(cofactor).
Species Streptococcus species
Protein Tag None
Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.