Learn More
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Name | Tyrosine Decarboxylase (S. faecalis) |
| Quantity | 25 U |
| Storage Requirements | Store at -20°C. |
| Regulatory Status | RUO |
| Common Name | TYROSINE |
| Biological Activity | ≥0.1 unit per mg dry weight. One Unit yields 1 umole of CO2 per minute from L-tyrosine at 37°C, pH 5.5. |
| Species | Streptococcus species |
| Protein Tag | None |
| Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.