Learn More
Description
Reconstitute with 1 mL of PBS buffer to yield a concentration of 100 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue, human U-87MG cell. IHC: rat brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | Tyrosine Hydroxylase |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | TH-16 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide |
| Gene | TH |
| Gene Accession No. | P04177, P07101, P24529 |
| Gene Alias | dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
