Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrphostin C15

Catalog No. aaj65227ma
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65227MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65227MA Supplier Thermo Scientific Chemicals Supplier No. J65227MA
Only null left
Add to Cart

CAS: 149092-50-2 | C19H15N3O3S2 | 397.47 g/mol

A selective ErbB2 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149092-50-2
Molecular Formula C19H15N3O3S2
Molecular Weight (g/mol) 397.47
MDL Number MFCD01074971
InChI Key KXDONFLNGBQLTN-UHFFFAOYSA-N
Synonym tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag
PubChem CID 6091659
ChEBI CHEBI:75405
IUPAC Name 3-{3-[(1,3-benzothiazol-2-ylsulfanyl)methyl]-4-hydroxy-5-methoxyphenyl}-2-cyanoprop-2-enamide
SMILES COC1=CC(C=C(C#N)C(N)=O)=CC(CSC2=NC3=CC=CC=C3S2)=C1O

Specifications

Color Yellow to Orange
Quantity 10 mg
Sensitivity Light sensitive
Formula Weight 397.47
Physical Form Powder
Chemical Name or Material Tyrphostin C15
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.