Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrphostin RG 13022 98.0+%, TCI America™
SDP

Catalog No. T3575100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3575100MG 100 mg
T357525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3575100MG Supplier TCI America Supplier No. T3575100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136831-48-6
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.3
MDL Number MFCD00236444
InChI Key DBGZNJVTHYFQJI-RIYZIHGNSA-N
Synonym 3-(3,4-Dimethoxyphenyl)-2-(pyridin-3-yl)acrylonitrile
PubChem CID 5468216
IUPAC Name (Z)-3-(3,4-dimethoxyphenyl)-2-pyridin-3-ylprop-2-enenitrile
SMILES COC1=C(C=C(C=C1)C=C(C#N)C2=CN=CC=C2)OC

Specifications

Melting Point 118°C
Color Yellow
Quantity 100 mg
Formula Weight 266.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tyrphostin RG 13022
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.