Learn More
DuPont™ Tyvek™ 400 SFR White Coverall ST120, Standard Fit Hood
Shop All DuPont Personal Protection ProductsDescription
- Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
- In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
- Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
- Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology.
- Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
- Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
- Relaxed fit coverall design features a diamond gusset crotch to allow for unrestricted movement and optimal fit when wearing primary flame-resistant protective clothing beneath
- Red trim elements - red external stitched seams, red zipper, and red chest patch border
- Patent-pending design
- Attached standard-fit hood with elastic around face opening
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Storm flap
Specifications
Specifications
| Product Type | Flame-resistant Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Disposable | Reusable |
| Waist Style | Standard |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.