Description
DuPont™ Tyvek™ 400 D pants have been designed for very specific applications where demanding comfort requirements are combined with limited protective requirements for frontal exposures. They are well suited for workers who are involved in a variety of strenuous activities that can lead to heat stress in applications that include:
- Wind Turbines
- Composites manufacturing
- Boat manufacturing
- Remediation
- Utilities
- Maintenance
- Glass manufacturing
Specifications
Specifications
| Size | 2X-Large |
| Material | Spunbond Polypropylene |
| Color | White |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.