Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 800 Hooded Coveralls

Catalog No. 19088820 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Large
Medium
Small
X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-820 6X-Large
19-715-365 Small
19-715-361 Medium
19-715-360 Large
19-715-359 X-Large
19-715-356 2X-Large
19-715-347 3X-Large
19-088-821 7X-Large
19-088-819 5X-Large
19-088-818 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Does not provide protection from ionizing radiation
Catalog No. 19-088-820 Supplier DuPont™ Supplier No. TJ198TWH6X0025PI
Add to Cart
Add to Cart
This item is not returnable. View return policy
Does not provide protection from ionizing radiation

Providing a suitable protective barrier against many low-concentration, water-based, inorganic chemicals and particles, down to one micron in size, Tyvek 800 coveralls offer an optimum fit with full freedom of movement.

Serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency.

  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604)
  • EN 1073-2 (protection against radioactive contamination)
  • Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals
  • Serged and over-taped seams for protection and strength
  • Self-adhesive chin flap for tight seal of suit to the mask
  • Elastic face, wrists and ankles as well as glued-in waist elastic
  • Elastic thumb loops keep sleeves in place
  • Distinct orange taping of the garment seams providing
  • color contrast

Order Info

25 per case

Compliance

CE certified.

Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B

TRUSTED_SUSTAINABILITY

Specifications

Size 6X-Large
Gender Unisex
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Liquid, Particle
Latex Free No
No. of Pockets None
For Use With (Application) Industrial cleaning, Petrochemical installations, sewers. Maintenance
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.