Description
Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible, yet provides superior contamination control and particulate filtration, unsurpassed liquid and particulate shielding and two-way contamination protection. Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint. Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags. ISO Class 5 (formerly Class 100) compatible. Constructed to hold up and keep you comfortable. Generous sizing permits freedom of movement. Plastic snaps won't rust or flake during processing. Available in bulk (no special cleaning), clean (ISO Class 4 [formerly Class 10] cleanroom processed), sterile (gamma irradiated), or clean and sterile. Every lot is 100% traceable.
- Excellent barrier to dry particulates, nonhazardous liquids, and microorganisms
- Bound neck with adjustable snaps Five snap front Tunneled elastic wrist Made from DuPont Tyvek™ brand protective material Unique, patented flashspinning process creates an excellent barrier to dry particles, microorganisms and nonhazardous liquids Comfortable, lightweight and durable Full traceability on all sterilized apparel Recyclable
Specifications
Specifications
| Size | X-Large |
| Material | Tyvek |
| Color | White |
| Sterility | Non-sterile |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.