Learn More
Description
- Made from Tyvek™ material, which is a patented flashspun high-density polyethylene, these boot covers provide an inherent barrier to particles, microorganisms, and non-hazardous liquid splashes.
- The fabric is specially processed to be exceptionally low-linting and to minimize particle shedding, which is a critical feature for maintaining the integrity of ISO Class 5-8 cleanrooms.
- Tyvek™ is known for its durability. The material has a high abrasion resistance and is strong enough to withstand rigorous use without wearing away, ensuring reliable protection.
- The boot covers are available in both clean-processed and sterile options.
- The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6, making them suitable for GMP Class A/B (ISO Class 5) environments.
- They are designed with features like tunnelled elastication at the shin and ankle, along with ankle ties, to provide a secure fit over footwear and prevent the boot cover from slipping or riding up.
- The boot covers are often folded to facilitate aseptic donning, minimizing the risk of contamination during the gowning process.
Specifications
Specifications
| Size | Large |
| Material | Tyvek |
| Latex Free | Yes |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Top |
| Sole Type | PVC |
| Thread Color | White |
| Packaging Quantity | 300 Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
