Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Hood
SDP
Catalog No. 17987879 Shop All New Pig Corporation Products
Catalog No. 17-987-879 Supplier New Pig Supplier No. WPL1017
Add to Cart
Add to Cart
Hood provides an excellent barrier to particles, microorganisms and non-hazardous light liquid splash in ISO Class 5-8 cleanrooms
  • Tyvek™ IsoClean™ hood provides an excellent, inherent barrier against dry particulates, microorganisms, and non-hazardous liquid splashes.
  • This is achieved through the use of flashspun high-density polyethylene, which is a key component of all Tyvek™ products.
  • The fabric is specially processed and treated to minimize the shedding of lint and particles, making it ideal for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • The material is known for its high strength and durability, with a strong resistance to tearing and abrasion, ensuring reliable protection even during rigorous use.
  • Tyvek™ unique nonwoven structure allows air and moisture vapor to pass through, which helps to reduce heat stress and keeps the wearer more comfortable, especially during long shifts.
  • The Tyvek™ IsoClean™ line includes a wide range of products, such as frocks, lab coats, boot covers, sleeve and hoods, which can be combined to create a complete head-to-toe ensemble for full-body protection.
  • A hallmark of the IsoClean™ line is the availability of both clean-processed and sterile options.
  • The sterile products are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for the most critical sterile applications, such as those in the pharmaceutical and life sciences industries.