Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 255 Coveralls
GREENER_CHOICE

Catalog No. 18888106 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-106 4X-Large
18-888-101 Medium
18-888-102 Large
18-888-103 X-Large
18-888-104 2X-Large
18-888-105 3X-Large
18-888-107 5X-Large
18-888-108 6X-Large
18-888-109 7X-Large
18-888-100 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 18-888-106 Supplier DuPont™ Supplier No. IC255BWH4X00250B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tyvek coveralls provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Tyvek™ flashspun polyolefin protective material
  • Patented flash-spinning process
  • Creates barrier to dry particles, microorganisms, non-hazardous liquids
  • Lightweight, durable
  • Serged seams with covered elastic options
  • Traceable
  • Gripper™ soled shoes for slip resistance
  • Bound seams and neck
  • Dolman sleeve
  • Covered elastic wrist, ankle
  • Thumb loop
  • Storm flap
  • Zipper closure
  • White in color
  • Bulk packaged
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large
Gender Unisex
Material Tyvek Flashspun Polyolefin
Color White
Disposable Single-use
Ankle Style Covered Elastic
Hood Type Standard
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Certifications/Compliance CE
Sleeve Style Dolman
For Use With (Application) Protective Barrier to Dry Particles and Microorganisms
Packaging Bulk
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.